accessibility menu, dialog, popup

UserName

CH3OOSCl

CAS RN 26555-40-8

IUPAC Name: (chlorosulfanyl)(methoxy)methanone
Synonyms: (chlorosulfanyl)(methoxy)methanone
Molecular Weight (g/mol): 126.55
Molecular Formula: C2H3ClO2S
InChi Key: TXJXPZVVSLAQOQ-UHFFFAOYSA-N
SMILES: COC(=O)SCl
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)