CAS RN 2648862-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diclofenac Levomenthol Ester, TRC
CAS: 2648862-09-1 Molecular Formula: C24 H29 Cl2 N O2 Molecular Weight (g/mol): 434.399 Synonym: (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2-(2-((2,6-Dichlorophenyl)amino)phenyl)acetate IUPAC Name: [(1R,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2-[2-(2,6-dichloroanilino)phenyl]acetate SMILES: CC(C)[C@H]1CC[C@@H](C)C[C@H]1OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl