accessibility menu, dialog, popup

UserName

CAS RN 261952-09-4

IUPAC Name: 3-methyl-5-(trifluoromethyl)benzoyl chloride
Synonyms: 3-methyl-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 222.59
Molecular Formula: C9H6ClF3O
InChi Key: STYLTOJEHPKJDF-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=C1)C(Cl)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results