accessibility menu, dialog, popup

UserName

CH3FFFN

CAS RN 261952-06-1

IUPAC Name: 4-methyl-3-(trifluoromethyl)benzonitrile
Synonyms: 4-methyl-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: YWAGJCPTOFPGIS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C(F)(F)F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results