accessibility menu, dialog, popup

UserName

CAS RN 261943-97-9

IUPAC Name: 2-(2,6-difluoro-3-methylphenyl)acetic acid
Synonyms: (2,6-difluoro-3-methylphenyl)acetic acid
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: XMNMLOQKAUVILT-UHFFFAOYSA-N
SMILES: CC1=C(F)C(CC(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results