accessibility menu, dialog, popup

UserName

CAS RN 261763-39-7

IUPAC Name: 2,6-difluoro-3-methylbenzoyl chloride
Synonyms: 2,6-difluoro-3-methylbenzoyl chloride
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: LXCIIPPXRHOCQN-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(C(Cl)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results