accessibility menu, dialog, popup

UserName

CAS RN 2592-19-0

H3CH3CCH3OOHN(S)HNOOCH3H3CCH3OONOO

CAS RN 2592-19-0

IUPAC Name: 4-nitrophenyl 2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoate
Synonyms: 4-nitrophenyl 2,6-bis[(tert-butoxycarbonyl)amino]hexanoate
Molecular Weight (g/mol): 467.52
Molecular Formula: C22H33N3O8
InChi Key: LYUXBTAUKJETMS-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCCC(NC(=O)OC(C)(C)C)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results