accessibility menu, dialog, popup

UserName

CAS RN 2586-93-8

ClH2NSH2N

CAS RN 2586-93-8

IUPAC Name: 2-amino-4-chlorobenzene-1-carbothioamide
Synonyms: 2-amino-4-chlorobenzenecarbothioamide
Molecular Weight (g/mol): 186.66
Molecular Formula: C7H7ClN2S
InChi Key: LTKVCPVQNOQNKY-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(N)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results