accessibility menu, dialog, popup

UserName

CAS RN 247170-19-0

CH3ONHFFFCl

CAS RN 247170-19-0

IUPAC Name: N-[2-chloro-4-(trifluoromethyl)phenyl]acetamide
Synonyms: N-[2-chloro-4-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 237.61
Molecular Formula: C9H7ClF3NO
InChi Key: QRMZQPODCUCPQD-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(Cl)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results