accessibility menu, dialog, popup

UserName

CH3OOO(S)O(S)O(R)OH(S)OH(S)OH(R)HO(S)(S)OH

CAS RN 24512-63-8

IUPAC Name: methyl (1S,4aS,7aS)-7-(hydroxymethyl)-1-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate
Synonyms: geniposide jasminoidin
Molecular Weight (g/mol): 388.37
Molecular Formula: C17H24O10
InChi Key: IBFYXTRXDNAPMM-BVTMAQQCSA-N
SMILES: COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]2[C@@H]1CC=C2CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results