CAS RN 24512-62-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gardenoside, TRC
CAS: 24512-62-7 Molecular Formula: C17 H24 O11 Molecular Weight (g/mol): 404.36 Synonym: (1S,4aS,7S,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,7,7a-tetrahydro-7-hydroxy-7-(hydroxymethyl)-cyclopenta[c]pyran-4-carboxylic acid methyl ester IUPAC Name: methyl (1S,4aS,7S,7aS)-7-hydroxy-7-(hydroxymethyl)-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-4a,7a-dihydro-1H-cyclopenta[c]pyran-4-carboxylate SMILES: COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3[C@@H]1C=C[C@@]3(O)CO