CAS RN 24502-79-2
IUPAC Name:
1-(5,8-dihydroxy-1,4-dioxo-1,4-dihydronaphthalen-2-yl)-4-methylpent-3-en-1-yl 3-methylbut-2-enoate
Synonyms:
1-(5,8-dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-en-1-yl 3-methylbut-2-enoate
Molecular Weight (g/mol):
370.4
Molecular Formula:
C21H22O6
InChi Key:
BATBOVZTQBLKIL-UHFFFAOYNA-N
SMILES:
CC(C)=CCC(OC(=O)C=C(C)C)C1=CC(=O)C2=C(O)C=CC(O)=C2C1=O
1
–
1
of
1
results
1
(Beta, Beta-Dimethylacryl)shikonin, TRC
CAS: 24502-79-2 Molecular Formula: C21H22O6 Molecular Weight (g/mol): 370.4 IUPAC Name: [(1R)-1-(5,8-dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-enyl] 3-methylbut-2-enoate SMILES: CC(=CC[C@@H](OC(=O)C=C(C)C)C1=CC(=O)c2c(O)ccc(O)c2C1=O)C