CAS RN 244193-56-4
CAS RN 244193-56-4
IUPAC Name:
1-decyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms:
1-decyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol):
310.19
Molecular Formula:
C14H27BF4N2
InChi Key:
QGUMDWFYJYXDTH-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CCCCCCCCCCN1C=C[N+](C)=C1
1
–
1
of
1
results
1
1-n-Decyl-3-methylimidazolium tetrafluoroborate, 97%
CAS: 244193-56-4 Molecular Formula: C14H27BF4N2 Molecular Weight (g/mol): 310.19 MDL Number: MFCD09038853 InChI Key: QGUMDWFYJYXDTH-UHFFFAOYSA-N Synonym: 1-decyl-3-methylimidazolium tetrafluoroborate,dsstox_cid_27891,dsstox_rid_82644,dsstox_gsid_47915,ksc200g6n,c10mim bf4,1-decyl-3-me-thylimidazolium tetrafluoroborate PubChem CID: 11461044 IUPAC Name: 1-decyl-3-methylimidazol-3-ium;tetrafluoroborate SMILES: F[B-](F)(F)F.CCCCCCCCCCN1C=C[N+](C)=C1