accessibility menu, dialog, popup

UserName

CNF

CAS RN 24075-34-1

IUPAC Name: 1-fluoro-4-isocyanobenzene
Synonyms: 1-fluoro-4-isocyanobenzene
Molecular Weight (g/mol): 121.11
Molecular Formula: C7H4FN
InChi Key: FEUHGXVGNQTPGQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)[N+]#[C-]
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results