CAS RN 2402-99-5
IUPAC Name:
3,5-dibromopyridin-1-ium-1-olate
Synonyms:
3,5-dibromopyridin-1-ium-1-olate
Molecular Weight (g/mol):
252.89
Molecular Formula:
C5H3Br2NO
InChi Key:
QBUQQUOGXPOBAQ-UHFFFAOYSA-N
SMILES:
[O-][N+]1=CC(Br)=CC(Br)=C1
1
–
1
of
1
results
1
3,5-Dibromopyridine-n-oxide, 97%, Thermo Scientific™
CAS: 2402-99-5 MDL Number: MFCD00160750 InChI Key: QBUQQUOGXPOBAQ-UHFFFAOYSA-N Synonym: 3,5-dibromopyridine 1-oxide,3,5-dibromopyridine n-oxide,3,5-dibromopyridine-n-oxide,3,5-dibromopyridin-1-ium-1-olate,3,5-dibromopyridine1-oxide,3,5-dibromopyridine-1-oxide,pyridine, 3,5-dibromo-,1-oxide,pubchem6616,3,5-dibromopyridin-1-ol,3,5-dibromopyridine n oxide PubChem CID: 620003 IUPAC Name: 3,5-dibromo-1-oxidopyridin-1-ium SMILES: C1=C(C=[N+](C=C1Br)[O-])Br