CAS RN 2367-73-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Dihydrotriamcinolone-16,17-acetonide-21-mesylate, TRC
CAS: 2367-73-9 Molecular Formula: C25H35FO8S Molecular Weight (g/mol): 514.6 Synonym: (11beta,16alpha)-9-Fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-21-[(methylsulfonyl)oxy]-pregn-4-ene-3,20-dione SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)COS(=O)(=O)C