accessibility menu, dialog, popup

UserName

CAS RN 23530-48-5

CH3H3CNHSOONOO

CAS RN 23530-48-5

IUPAC Name: 4-nitro-N-(propan-2-yl)benzene-1-sulfonamide
Synonyms: N-isopropyl-4-nitrobenzenesulfonamide
Molecular Weight (g/mol): 244.27
Molecular Formula: C9H12N2O4S
InChi Key: IMRZQROLAZHXHR-UHFFFAOYSA-N
SMILES: CC(C)NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results