CAS RN 234111-08-1
IUPAC Name:
2-bromo-6-iodopyridine
Synonyms:
2-bromo-6-iodopyridine
Molecular Weight (g/mol):
283.89
Molecular Formula:
C5H3BrIN
InChi Key:
KJOQMWRTNVBXEV-UHFFFAOYSA-N
SMILES:
BrC1=NC(I)=CC=C1
1
–
1
of
1
results
1
2-Bromo-6-iodopyridine, 97%
CAS: 234111-08-1 Molecular Formula: C5H3BrIN Molecular Weight (g/mol): 283.89 MDL Number: MFCD08059557 InChI Key: KJOQMWRTNVBXEV-UHFFFAOYSA-N Synonym: pyridine, 2-bromo-6-iodo,2-bromo-6-iodo-pyridine,6-bromo-2-iodopyridine,pyridine,2-bromo-6-iodo PubChem CID: 11312130 IUPAC Name: 2-bromo-6-iodopyridine SMILES: BrC1=NC(I)=CC=C1