CAS RN 2331211-44-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Emtricitabine 6’-Disulfide, TRC
CAS: 2331211-44-8 Molecular Formula: C16 H18 F2 N6 O4 S4 Molecular Weight (g/mol): 524.609 Synonym: (-)-2’,3’-Dideoxy-5-fluoro-3’-thiacytidine 6’-Disulfide,(-)-2'-Deoxy-5-fluoro-3'-thiacytidine 6’-Disulfide IUPAC Name: 4-amino-1-[(2R,5S)-2-[[[(2R,5S)-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-1,3-oxathiolan-2-yl]methyldisulfanyl]methyl]-1,3-oxathiolan-5-yl]-5-fluoropyrimidin-2-one SMILES: NC1=NC(=O)N(C=C1F)[C@@H]2CS[C@H](CSSC[C@@H]3O[C@@H](CS3)N4C=C(F)C(=NC4=O)N)O2