accessibility menu, dialog, popup

UserName

CAS RN 22812-90-4

H3CSOH

CAS RN 22812-90-4

IUPAC Name: 2-(propylsulfanyl)ethan-1-ol
Synonyms: 2-(propylsulfanyl)ethanol
Molecular Weight (g/mol): 120.21
Molecular Formula: C5H12OS
InChi Key: KCWWXXYQPUDKBX-UHFFFAOYSA-N
SMILES: CCCSCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results