accessibility menu, dialog, popup

UserName

CAS RN 2277-19-2

IUPAC Name: (6Z)-non-6-enal
Synonyms: cis-6-non-6-enal
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: RTNPCOBSXBGDMO-ARJAWSKDSA-N
SMILES: CC\C=C/CCCCC=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results