accessibility menu, dialog, popup

UserName

CAS RN 22767-90-4

IUPAC Name: 1,1,1-trifluoro-5,5-dimethylhexane-2,4-dione
Synonyms: 1,1,1-trifluoro-5,5-dimethylhexane-2,4-dione
Molecular Weight (g/mol): 196.17
Molecular Formula: C8H11F3O2
InChi Key: BVPKYBMUQDZTJH-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CC(=O)C(F)(F)F
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results