accessibility menu, dialog, popup

UserName

H3CONClN

CAS RN 22536-63-6

IUPAC Name: 2-chloro-4-methoxypyrimidine
Synonyms: 2-chloro-4-methoxypyrimidine
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: BDXYNMVQMBCTDB-UHFFFAOYSA-N
SMILES: COC1=CC=NC(Cl)=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results