accessibility menu, dialog, popup

UserName

CAS RN 22482-43-5

ONOClCl(E)

CAS RN 22482-43-5

IUPAC Name: 1,3-dichloro-2-[(1E)-2-nitroethenyl]benzene
Synonyms: 1,3-dichloro-2-[(1E)-2-nitroethenyl]benzene
Molecular Weight (g/mol): 218.03
Molecular Formula: C8H5Cl2NO2
InChi Key: VXNHQIKJDIOBEC-SNAWJCMRSA-N
SMILES: [O-][N+](=O)\C=C\C1=C(Cl)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results