accessibility menu, dialog, popup

UserName

CAS RN 2248-46-6

H3COOFF

CAS RN 2248-46-6

IUPAC Name: ethyl 2,2-difluoro-2-phenylacetate
Synonyms: ethyl 2,2-difluoro-2-phenylacetate
Molecular Weight (g/mol): 200.19
Molecular Formula: C10H10F2O2
InChi Key: KCMSDCHUELUJPX-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results