accessibility menu, dialog, popup

UserName

CH3H3CCH3PH3CH3CCH3

CAS RN 224311-51-7

IUPAC Name: {[1,1'-biphenyl]-2-yl}di-tert-butyl)phosphane
Synonyms: [1,1'-biphenyl]-2-yldi-tert-butyl)phosphane
Molecular Weight (g/mol): 298.41
Molecular Formula: C20H27P
InChi Key: CNXMDTWQWLGCPE-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C1=CC=CC=C1C1=CC=CC=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results