accessibility menu, dialog, popup

UserName

CAS RN 223655-16-1

FePt-But-Bu

CAS RN 223655-16-1

IUPAC Name: λ²-iron(2+) 1-(di-tert-butylphosphanyl)cyclopenta-2,4-dien-1-ide cyclopenta-2,4-dien-1-ide
Synonyms: λ²-iron(2+) 1-(di-tert-butylphosphanyl)cyclopenta-2,4-dien-1-ide cyclopentadienide
Molecular Weight (g/mol): 330.23
Molecular Formula: C18H27FeP
InChi Key: IVWFNMCAPPQZMP-UHFFFAOYSA-N
SMILES: [Fe++].[CH-]1C=CC=C1.CC(C)(C)P([C-]1C=CC=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results