accessibility menu, dialog, popup

UserName

CAS RN 22364-68-7

CH3N

CAS RN 22364-68-7

IUPAC Name: 2-(2-methylphenyl)acetonitrile
Synonyms: 2-methylbenzyl cyanide 2-methylbezeneacetonitrile
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: WMGVPDQNPUQRND-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CC#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results