accessibility menu, dialog, popup

UserName

CuCuS

CAS RN 22205-45-4

IUPAC Name: bis(λ¹-copper(1+)) λ⁴-sulfanediidyl
Synonyms: copper(I) sulfide
Molecular Weight (g/mol): 160.16
Molecular Formula: Cu2HS
InChi Key: SCQKXCATDJAJTA-UHFFFAOYSA-N
SMILES: [SH--].[Cu+].[Cu+]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results