CAS RN 22062-54-0
IUPAC Name:
(5-fluoro-2-methylphenyl)methanol
Synonyms:
(5-fluoro-2-methylphenyl)methanol
Molecular Weight (g/mol):
140.16
Molecular Formula:
C8H9FO
InChi Key:
WEXCPHVQJFWNFM-UHFFFAOYSA-N
SMILES:
CC1=C(CO)C=C(F)C=C1
1
–
1
of
1
results
1
5-Fluoro-2-methylbenzyl alcohol, 98%, Thermo Scientific™
CAS: 22062-54-0 Molecular Formula: C8H9FO Molecular Weight (g/mol): 140.157 MDL Number: MFCD01631527 InChI Key: WEXCPHVQJFWNFM-UHFFFAOYSA-N Synonym: 5-fluoro-2-methylbenzyl alcohol,5-fluoro-2-methylphenyl methanol,benzenemethanol,5-fluoro-2-methyl,3-fluoro-6-methylbenzyl alcohol,4-fluoro-2-hydroxymethyl toluene,5-fluoro-2-methylphenyl methan-1-ol PubChem CID: 2774595 IUPAC Name: (5-fluoro-2-methylphenyl)methanol SMILES: CC1=C(C=C(C=C1)F)CO