CAS RN 21901-34-8
IUPAC Name:
3-methyl-5-nitro-1,2-dihydropyridin-2-one
Synonyms:
3-methyl-5-nitro-1H-pyridin-2-one
Molecular Weight (g/mol):
154.13
Molecular Formula:
C6H6N2O3
InChi Key:
FPTYZBDNBMVYCL-UHFFFAOYSA-N
SMILES:
CC1=CC(=CNC1=O)[N+]([O-])=O
1
–
1
of
1
results
1
2-Hydroxy-3-methyl-5-nitropyridine, 97%, Thermo Scientific™
CAS: 21901-34-8 Molecular Formula: C6H6N2O3 Molecular Weight (g/mol): 154.125 MDL Number: MFCD03095073 InChI Key: FPTYZBDNBMVYCL-UHFFFAOYSA-N Synonym: 2-hydroxy-3-methyl-5-nitropyridine,3-methyl-5-nitropyridin-2-ol,2-hydroxy-5-nitro-3-picoline,3-methyl-5-nitro-2-pyridone,2-hydroxy-5-nitro-3-methylpyridine,2 1h-pyridinone, 3-methyl-5-nitro,3-methyl-5-nitro-2 1h-pyridinone,3-methyl-5-nitro-2-pyridinol,3-methyl-5-nitro-pyridin-2-ol,6-hydroxy-5-methyl-3-nitropyridine PubChem CID: 89097 IUPAC Name: 3-methyl-5-nitro-1H-pyridin-2-one SMILES: CC1=CC(=CNC1=O)[N+](=O)[O-]