CAS RN 21851-89-8
CAS RN 21851-89-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(diphenylphosphinomethyl)phenylphosphine oxide, 95%, Thermo Scientific™
CAS: 21851-89-8 Molecular Formula: C32H29O3P3 Molecular Weight (g/mol): 554.50 MDL Number: MFCD00482218 InChI Key: GPDBOGDYDXUTRD-UHFFFAOYSA-N Synonym: bis diphenylphosphinomethyl phenylphosphine oxide,diphenylphosphanyl methyl phenyl phosphoryl methyl diphenylphosphane,1,1,3,5,5-pentaphenyl-1,3,5-triphosphapentane trioxide IUPAC Name: [({[(diphenylphosphoroso)methyl](phenyl)phosphoryl}methyl)(phenyl)phosphoroso]benzene SMILES: O=P(CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)(CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1