accessibility menu, dialog, popup

UserName

CAS RN 2181-44-4

IUPAC Name: trimethylsulfanium methyl sulfate
Synonyms: trimethylsulfonium methyl sulfate(1-)
Molecular Weight (g/mol): 188.26
Molecular Formula: C4H12O4S2
InChi Key: ANXKZXRDXAZQJT-UHFFFAOYSA-M
SMILES: C[S+](C)C.COS([O-])(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results