accessibility menu, dialog, popup

UserName

CAS RN 21803-75-8

NH2ClN

CAS RN 21803-75-8

IUPAC Name: 4-amino-3-chlorobenzonitrile
Synonyms: 4-amino-3-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: OREVCMGFYSUYPX-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Cl)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results