CAS RN 21617-12-9
IUPAC Name:
4,8-dichloroquinoline
Synonyms:
4,8-dichloroquinoline
Molecular Weight (g/mol):
198.05
Molecular Formula:
C9H5Cl2N
InChi Key:
SDPCOMBBZFETLG-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC2=C(Cl)C=CN=C12
1
–
1
of
1
results
1
4,8-Dichloroquinoline, TRC
CAS: 21617-12-9 Molecular Formula: C9H5NCl2 Molecular Weight (g/mol): 198.05 Synonym: Hydroxychloroquine Impurity 6 SMILES: Clc1ccnc2c(Cl)cccc12