accessibility menu, dialog, popup

UserName

CAS RN 2140-76-3

CH3O(R)(R)OH(R)OHO(R)NOHNO

CAS RN 2140-76-3

IUPAC Name: 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 258.23
Molecular Formula: C10H14N2O6
InChi Key: SXUXMRMBWZCMEN-ZOQUXTDFSA-N
SMILES: CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results