CAS RN 2127839-79-4
CAS RN 2127839-79-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max
CAS: 2127839-79-4 Molecular Formula: H24Mo7N6O24 Molecular Weight (g/mol): 1163.85 MDL Number: MFCD00064633 InChI Key: MISTUIVWPMGXFX-UHFFFAOYSA-T Synonym: hexaammonium molybdate,ammoniummolybdatehydrate,nh4 6mo7o4 PubChem CID: 71306766 IUPAC Name: hexaazanium;dioxido(dioxo)molybdenum SMILES: [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11