accessibility menu, dialog, popup

UserName

CAS RN 211943-12-3

SBrNNHNN

CAS RN 211943-12-3

IUPAC Name: 5-(5-bromothiophen-2-yl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(5-bromothiophen-2-yl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 231.07
Molecular Formula: C5H3BrN4S
InChi Key: GJHHXCMGAACACG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=NNN=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results