accessibility menu, dialog, popup

UserName

NH2OFFFF

CAS RN 207986-20-7

IUPAC Name: 3-fluoro-5-(trifluoromethyl)benzamide
Synonyms: 3-fluoro-5-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 207.13
Molecular Formula: C8H5F4NO
InChi Key: BBUBHNZCRQRAQG-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(F)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results