accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNSOOFFFNHOOCH3CH3H3C

CAS RN 207857-15-6

IUPAC Name: tert-butyl N-({[(tert-butoxy)carbonyl]amino}(trifluoromethanesulfonylimino)methyl)carbamate
Synonyms: tert-butyl N-{[(tert-butoxycarbonyl)amino](trifluoromethanesulfonylimino)methyl}carbamate
Molecular Weight (g/mol): 391.36
Molecular Formula: C12H20F3N3O6S
InChi Key: GOQZIPJCBUYLIR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC(NC(=O)OC(C)(C)C)=NS(=O)(=O)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results