CAS RN 2076-91-7
CAS RN 2076-91-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TPT-260 Dihydrochloride, MedChemExpress
MedChemExpress TPT-260 Dihydrochloride (NSC55712) is a thiophene thiourea derivative with molecule weight 260.00 in free base form; There is no formal name yet, we temporally call this molecule as TPT-260.