accessibility menu, dialog, popup

UserName

CAS RN 2037-81-2

H3COOSNNHNOO

CAS RN 2037-81-2

IUPAC Name: ethyl 5-[(phenoxycarbonyl)amino]-1,2,3-thiadiazole-4-carboxylate
Synonyms: ethyl 5-[(phenoxycarbonyl)amino]-1,2,3-thiadiazole-4-carboxylate
Molecular Weight (g/mol): 293.3
Molecular Formula: C12H11N3O4S
InChi Key: FCPHNPRVMWSGSK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results