accessibility menu, dialog, popup

UserName

H3COOOONH

CAS RN 202195-67-3

IUPAC Name: methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate
Synonyms: methyl 3-oxo-2,4-dihydro-1,4-benzoxazine-6-carboxylate
Molecular Weight (g/mol): 207.19
Molecular Formula: C10H9NO4
InChi Key: WPGYGHFVLMZWHI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=C(OCC(=O)N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results