accessibility menu, dialog, popup

UserName

CAS RN 200875-76-9

ClClNHOCl

CAS RN 200875-76-9

IUPAC Name: 4-chloro-N-(2,3-dichlorophenyl)butanamide
Synonyms: 4-chloro-N-(2,3-dichlorophenyl)butanamide
Molecular Weight (g/mol): 266.55
Molecular Formula: C10H10Cl3NO
InChi Key: MCHSMQZMPJDCLZ-UHFFFAOYSA-N
SMILES: ClCCCC(=O)NC1=C(Cl)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results