accessibility menu, dialog, popup

UserName

FFFBr

CAS RN 1997-80-4

IUPAC Name: 1-(2-bromoethyl)-3-(trifluoromethyl)benzene
Synonyms: 1-(2-bromoethyl)-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 253.06
Molecular Formula: C9H8BrF3
InChi Key: AZLYAPKPZBXCGX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CCBr)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results