accessibility menu, dialog, popup

UserName

FOFFF

CAS RN 199292-40-5

IUPAC Name: [2-fluoro-5-(trifluoromethyl)phenyl](phenyl)methanone
Synonyms: [2-fluoro-5-(trifluoromethyl)phenyl](phenyl)methanone
Molecular Weight (g/mol): 268.21
Molecular Formula: C14H8F4O
InChi Key: SRXSIASKXYCRCC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1C(=O)C1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results