accessibility menu, dialog, popup

UserName

NO

CAS RN 198878-42-1

IUPAC Name: hydrate quinolin-1-ium-1-olate
Synonyms: quinoline-1-oxide hydrate
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: CUSWDTBBCIXCRB-UHFFFAOYSA-N
SMILES: O.[O-][N+]1=CC=CC2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results