CAS RN 198561-04-5
CAS RN 198561-04-5
IUPAC Name:
(2S)-3-(4-bromophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms:
(2S)-3-(4-bromophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol):
465.32
Molecular Formula:
C24H19BrNO4
InChi Key:
TVBAVBWXRDHONF-QFIPXVFZSA-M
SMILES:
[O-]C(=O)[C@H](CC1=CC=C(Br)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
4-Bromo-N-Fmoc-L-phenylalanine, 95%
CAS: 198561-04-5 Molecular Formula: C24H20BrNO4 Molecular Weight (g/mol): 466.33 MDL Number: MFCD00273460 InChI Key: TVBAVBWXRDHONF-QFIPXVFZSA-N Synonym: fmoc-l-4-bromophenylalanine,s-n-fmoc-4-bromophenylalanine,fmoc-phe 4-br-oh,fmoc-4-bromo-l-phenylalanine,fmoc-l-4-br-phe-oh,fmoc-p-bromo-phe-oh,fmoc-d-4-bromophenylalanine,fmoc-l-4-bromophe,fmoc-l-phe 4-br-oh,fmoc-4'-bromo-l-phe PubChem CID: 2734449 SMILES: OC(=O)[C@H](CC1=CC=C(Br)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12