CAS RN 19841-72-6
IUPAC Name:
2,2-dimethyloctan-3-ol
Synonyms:
2,2-dimethyloctan-3-ol
Molecular Weight (g/mol):
158.29
Molecular Formula:
C10H22O
InChi Key:
AZIWNYAWODSERA-UHFFFAOYNA-N
SMILES:
CCCCCC(O)C(C)(C)C
1
–
1
of
1
results
1
2,2-Dimethyl-3-octanol, 98%, Thermo Scientific™
CAS: 19841-72-6 Molecular Formula: C10H22O Molecular Weight (g/mol): 158.29 MDL Number: MFCD00021945 InChI Key: AZIWNYAWODSERA-UHFFFAOYNA-N PubChem CID: 140618 IUPAC Name: 2,2-dimethyloctan-3-ol SMILES: CCCCCC(O)C(C)(C)C