CAS RN 19523-57-0
IUPAC Name:
2-ethoxynaphthalene-1-carbaldehyde
Synonyms:
2-ethoxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol):
200.24
Molecular Formula:
C13H12O2
InChi Key:
IMNKQTWVJHODOS-UHFFFAOYSA-N
SMILES:
CCOC1=CC=C2C=CC=CC2=C1C=O
1
–
1
of
1
results
1
2-Ethoxy-1-naphthaldehyde, 98%, Thermo Scientific™
CAS: 19523-57-0 Molecular Formula: C13H12O2 Molecular Weight (g/mol): 200.24 MDL Number: MFCD00046427 InChI Key: IMNKQTWVJHODOS-UHFFFAOYSA-N Synonym: 2-ethoxy-1-naphthaldehyde,1-naphthalenecarboxaldehyde, 2-ethoxy,2-ethoxy-1-naphthalenecarboxaldehyde,1-naphthaldehyde, 2-ethoxy,1-naphthalenecarboxaldehyde,2-ethoxy,2-ethoxynaphthalenecarbaldehyde,2-athoxy-naphthal,acmc-1bs4w,2-ethoxy-1-naphthalenecarbaldehyde,2-ethoxy-naphthalene-1-carbaldehyde PubChem CID: 88099 IUPAC Name: 2-ethoxynaphthalene-1-carbaldehyde SMILES: CCOC1=CC=C2C=CC=CC2=C1C=O